Learn More
Invitrogen™ PSI-CLONE™ 96-Well PCR Purification Plates
Description
Applied Biosystems PSI-Clone™ PCR 96-Well plates are for the rapid purification of PCR reaction products using either vacuum- or centrifugation-based protocols. Both protocols are fast and result in high-yield, high-purity DNA ideal for molecular biology experiments.
DNA and RNA Purification and Analysis, DNA Extraction, PCR Product Clean-Up
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Format | 96-well Plate |
| Quantity | 50 Plates |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.