missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Plant RNA Isolation Aid

Product Code. 10773837
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10773837 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10773837 Supplier Invitrogen™ Supplier No. AM9690
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recommended for use with the RNAqueous Kit for purification of total RNA from plant tissues. One bottle of 10 mL Invitrogen reagent is provided.

Recommended for use with the RNAqueous Kit for purification of total RNA from plant tissues. One bottle of 10 mL Invitrogen reagent is provided.

Invitrogen's Plant RNA Isolation Aid contains polyvinylpyrrolidone (PVP), a high molecular weight polymer that binds to contaminants such as polyphenolics and polysaccharides that are commonly present in plant tissues. Glass fiber filter–based methods of isolation, such as the RNAqueous Kit, are especially suited for plant RNA isolation because it does not include any alcohol precipitation steps; many common contaminants found in plant tissue are known to co-precipitate with nucleic acid during alcohol precipitation. When used with the RNAqueous Kit, the procedure consists of disrupting the plant tissue in a guanidinium-based lysis solution to which the Plant RNA Isolation Aid has been added. A brief centrifugation then removes the contaminants. The supernatant is passed through a glass fiber filter which binds RNA. The filter is then washed and the RNA is eluted in a small volume (50μL or more) of elution solution.

• Removes common plant contaminants such as polyphenolics and polysaccharides
• No phenol extraction, alcohol precipitation, or proteinase digestion steps required when used with the RNAqueous Kit
• Not high-throughput compatible (Manual)

Accessory Product
The Plant RNA Isolation Aid is designed for use with the RNAqueous Kit (Cat. No. AM1912) for RNA purification from plant tissues.

Applications
DNA and RNA Purification and Analysis, RNA Extraction, Total RNA Isolation, Total RNA from Plant Cells

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store Plant RNA Isolation Aid at room temperature or at 4°C.
Isolation Technology PVP Reagent
Sample Type Plant Samples
Final Product Type RNA Supernatant (for use with RNAqueous™ kit)
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Equipment) RNAqueous™ Kit
Quantity 10 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.