Learn More
Invitrogen™ Phenol:Chloroform:IAA, 25:24:1, pH 6.6
Description
Ambion Acid Phenol:Chloroform:IAA (25:24:1) is premixed and supplied at pH 6.6 (pH 7.9 ± 0.2), with included buffer. Acid Phenol:Chloroform:IAA is provided in one bottle of 400 mL. In RNA and DNA extraction procedures, Acid Phenol:Chloroform:IAA (pH 7.9) helps to stabilize the interface and prevents foaming when mixing. Preparation of phenol for use in molecular biology applications is a time-consuming and often hazardous procedure due to its toxic and corrosive nature. Ambion premixed, quality tested, saturated phenols are ready-to-use, eliminating handling problems and offering a more convenient, safer, and easier alternative to preparing solutions from crystalline phenol.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Chemical Name or Material | Phenols |
| Content And Storage | Store at 4°C or -20°C. |
| Packaging Type | Bottle |
| pH | 6.6 |
| Product Line | Ambion |
| Purity | Molecular Biology Grade |
| Quantity | 400 mL |
| Shipping Condition | Room Temperature |
| Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.