Learn More
Invitrogen™ NorthernMax™ Kit
Product Code. 10574825
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
Product Code. Quantity unitSize
10574825 1 kit Each
1 options
For Research Use Only. All usage must comply with product instructions.
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10574825 Supplier Invitrogen™ Supplier No. AM1940
For Research Use Only. All usage must comply with product instructions.
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Includes
Positive Control RNA, pTRI-GAPDH Mouse, DECAtemplate -actin-Mouse, Formaldehyde Load Dye, ULTRAhyb Buffer, 10X Denaturing Gel Buffer, and 10X MOPS Gel Running Buffer, Agarose-LE, Transfer Buffer, Low Stringency Wash Solution No. 1, High Stringency Wash Solution No. 2, and RnaseZap™, nuclease-free water
A complete set of RNase-free reagents for running formaldehyde-based northern analysis
  • Suitable for the investigator who wants to use familiar reagents while enjoying high sensitivity
  • Sufficient reagents are provided to process 1000cm2 membrane
  • Complete kit contains sufficient buffer, gel and wash reagents for 10 to 20 gels
  • The included ULTRAhyb™ Ultrasensitive Hybridization Solution increases the sensitivity of RNA probes up to 20x and DNA probes up to 100x
  • Hybridization can be completed in 2 hours for many messages
  • Compatible with DNA, RNA, or oligo probes labeled isotopically or nonisotopically
  • Compatible with formaldehyde-containing gels
  • An RNA sample and template for probe synthesis are also provided as controls for each kit
DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage • Positive Control RNA, pTRI-GAPDH Mouse, DECAtemplate §-actin-Mouse, and Formaldehyde Load Dye should all be stored at -20°C.
• ULTRAhyb™ Buffer, 10X Denaturing Gel Buffer, and 10X MOPS Gel Running Buffer should be stored at 4°C.
• Agarose-LE™, Transfer Buffer, Low Stringency Wash Solution #1, High Stringency Wash Solution #2, and RNaseZap™ should be stored at room temperature.
• Nuclease-free water may be stored at any temperature.
Gel Compatibility Formaldehyde-Containing Gels
Quantity 1 kit
Shipping Condition Room Temperature
Product Type Northern Analysis Reagents