missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse RiboPure™-Blood RNA Isolation Kit with Manual

Product Code. p-4918751
Click to view available options
Quantity:
25 preps
Product Type:
Kit
Kit with manual
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10331325

Brand: Invitrogen™ AM1951

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Acid–Phenol:Chloroform, 3M Sodium Acetate pH 5.5, Wash Solution 1 Concentrate, Wash Solution 2/3 Concentrate, 2mL tubes pre-loaded with RNAlater, Lysis Solution, Filter Cartridges, Collection Tubes, Nuclease–free Water and Elution solution

For the purification of extremely high-quality total RNA (and optionally, microRNA) from mouse or rat blood in about 35 minutes.

For the purification of extremely high-quality total RNA (and optionally, microRNA) from mouse or rat blood in about 35 minutes. The Ambion™ kit includes sufficient reagents for 25 RNA purifications from whole blood samples ranging from 0.1–0.5 each.

• Includes RNAlater™, predispensed in 2 mL tubes, for RNA stabilization
• Resulting RNA is suitable for stringent downstream applications such as array analysis and qRT-PCR

Due to interference from the enzymatic inhibitors and anticoagulants present in blood samples, RNA purification from whole blood requires a stringent isolation method that yields high-quality RNA depleted of inhibiting agents and contaminants. The kit includes microfuge tubes preloaded with RNAlater™ Tissue Collection:RNA Stabilization Solution, an aqueous, nontoxic storage reagent that prevents RNA degradation and preserves RNA profiles within samples. Stabilized samples can be stored at room temperature for up to 3 days or at –20°C for long-term storage. The related Figures show the performance of RNA isolated with the Mouse RiboPure™-Blood Kit in real-time RT-PCR. The kit includes an alternative protocol for isolating total RNA that contains microRNA. Typically, 30–55 μg of total RNA (containing microRNA) is obtained from 0.5 mL mouse blood. Yields from rat blood are considerably more variable, generally 18–40 μg from 0.5 mL blood.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Acid-Phenol:Chloroform, 3M Sodium Acetate pH 5.5, Wash Solution 1 Concentrate, Wash Solution 2/3 Concentrate at 4°C. Store 2 mL tubes pre-loaded with RNAlater, Lysis Solution, Filter Cartridges, Collection Tubes, Nuclease-free Water and Elution solution at room temperature.
Format Filter Cartridge
Isolation Technology Organic Extraction, Spin Column
Purification Time 35 min.
Species Mouse
Sample Type Blood
Final Product Type Total RNA, micro RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Microarray Analysis, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Northern Blotting
Includes Acid–Phenol:Chloroform, 3M sodium acetate pH 5.5, wash solution 1 concentrate, wash solution 2/3 concentrate, 2mL tubes pre-loaded with RNAlater, lysis solution, filter cartridges, collection tubes, nuclease-free water and elution solution
Prep Scale 100 μL to 1 mL Blood Samples (Medium-scale)
Product Line Ambion, RiboPure
Product Type Kit
Quantity 25 preps
Shipping Condition Room Temperature
Starting Material Amount Up to 0.5 ml whole blood
Target miRNA, Total RNA
Yield 40 μg
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.