missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ambion™ mMESSAGE mMACHINE™ SP6 Transcription Kit

Product Code. 10086184
Change view
Click to view available options
Quantity:
25 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10086184 25 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10086184 Supplier Invitrogen™ Supplier No. AM1340M

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

For Use With (Application) Gene Expression Analysis and Genotyping, in vitro Transcription
Includes SP6 Enzyme Mix, 10X Reaction Buffer, 2XNTP/CAP Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, Gel Loading Buffer II, and Nuclease-free Water
No. of Reactions 25
Product Type Kit with manual
Quantity 25 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.