missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mirVana™ miRNA Detection Kit

Product Code. 10499014
Change view
Click to view available options
Quantity:
100 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10499014 100 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10499014 Supplier Invitrogen™ Supplier No. AM1552

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Hybridization Buffer, A/T1 Solution, Yeast RNA (5mg/mL), RNase Digestion Buffer, RNase Inactivation/PPT Solution, Loading Buffer II, Mouse Kidney Total RNA, Control Template (10μM), Probe Elution Buffer and Nuclease-free Water

Provides a fast and sensitive method for detecting small RNAs

The assay is 100-500 times more sensitive than Northern analysis, it is able to detect as little as 10 attomoles (10-17 mol) of target RNA.

Rapid Procedure Based on Solution Hybridization
The mirVana™ miRNA Detection Kit is based on a simple solution hybridization principle where the RNA sample is simply mixed with radiolabeled RNA probe(s) and hybridized. Unhybridized RNA and excess probe are removed by a digestion step and the hybridized, protected RNA fragments are then recovered using a patented single-step technology for simultaneous ribonuclease inactivation and nucleic acid precipitation. RNA samples are then resuspended and analyzed on a denaturing polyacrylamide gel.

Simultaneous Detection of siRNA Expression and Target Gene Knockdown
Because multiple RNA species of the same or differing size can be detected in the same sample, the mirVana miRNA Detection Kit is ideal for correlating siRNA expression levels with target mRNA knockdown. An example of this application demonstrates that GAPDH mRNA levels were reduced in cells expressing a GAPDH siRNA, but not in cells expressing a control siRNA.

Accessory Products:
The mirVana miRNA Probe Construction Kit (Cat. No. AM1550) is available for the preparation and labeling of probes by in vitro transcription and the mirVana™ Probe & Marker Kit is available for 5' end labeling of oligonucleotide probes and the included Decade Markers (Cat. No. AM1554).

  • Includes sufficient reagents for 100 reactions
  • Sensitive: detect miRNA or siRNA in as little as 50ng total RNA
  • Specific: extremely low backgrounds
  • Simple and fast: single-tube procedure eliminates the need to transfer to membrane for hybridization
  • Multiple target detection: detect multiple small RNAs and mRNAs in the same sample

Hybridization-Based miRNA Detection, RNAi, RNAi, Epigenetics and Non-Coding RNA Research, miRNA and Non-Coding RNA Analysis, siRNA

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Kit, miRNA Detection
Detection Method Radioactive
Content And Storage Hybridization Buffer, A⁄T1 Solution, Yeast RNA (5 mg⁄mL), RNase Digestion Buffer,
RNase Inactivation⁄PPT Solution, Loading Buffer II, Mouse Kidney Total RNA, Control Template (10 μM), and Probe Elution Buffer are all stored at –20°C. Nuclease-free Water can be stored at any temperature.
Sample Type miRNA
Shipping Condition Dry Ice
Product Line mirVana, Ambion
For Use With (Application) Solution Hybridization
Quantity 100 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.