missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MessageAmp™ Premier RNA Amplification Kit

Product Code. 10534445
Change view
Click to view available options
Quantity:
10 Reactions
100 Reactions
30 Reactions
Unit Size:
1 set
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10534445 100 Reactions 1 set
10668255 10 Reactions 1 set
10126824 30 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10534445 Supplier Invitrogen™ Supplier No. 4383452

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

110μL First Strand Enzyme Mix; 440μL First Strand Buffer Mix; 220μL Second Strand Enzyme Mix; 550μL Second Strand Buffer Mix; 660μL T7 Enzyme Mix); 2 x 1.1mL T7 Biotin IVT Mix; 10μL Control RNA 1mg/mL; 1.75mL Nuclease-free Water; 6mL aRNA Binding Buffer Concentrate; 1.2mL RNA Binding Beads; 30mL aRNA Wash Solution Concentrate; 15mL aRNA Elution Solution; 1mL 5X Array Fragmentation Buffer; 2 U-Bottom Plates 4 PCR Plates; 1 Reservoir; 10mL Nuclease-free Water

Innovative kit for the preparation of RNA samples for microarray analysis

Kit employs the proven methodology for RNA sample preparation and labeling based on the original T7 in vitro transcription (IVT) amplification technology, known as the Eberwine or reverse transcription-IVT (RT-IVT) method. The RT-IVT method is considered the gold standard for sample preparation in microarray-based expression profiling. It is well documented in the current scientific literature and was experimentally validated using TaqMan™ RT-PCR. Builds upon the linear RNA amplification method and reagents developed for MessageAmp™ II Kits with enhancements that create a simplified workflow, improved performance, and more flexible RNA input requirements.

Using the MessageAmp Premier RNA Amplification Kit
Based on this proven enzymatic amplification and labeling strategy, the MessageAmp Premier procedure starts with a simple cDNA synthesis reaction using ArrayScript™ reverse transcriptase. No cleanup step is required after the RT; cDNA is used directly in a high yield IVT reaction using MEGAscript technology. The IVT is configured to incorporate the modified nucleotide, biotin-UTP, into the aRNA synthesized. (In this protocol, the amplified RNA is referred to as aRNA; in the literature, it is also commonly called cRNA.) Once purified, the aRNA is suitable for use on microarray gene expression analysis platforms designed for biotin-modified antisense RNA samples.

  • Very flexible RNA input requirements: 20-500ng total RNA, depending on the tissue or cell type
  • Streamlined workflow: depending on the RNA input amounts, only a single day of processing time is required to complete a microarray experiment
  • Simplified protocol and streamlined reagents
  • Complete kit: everything needed to prepare biotin-modified aRNA is included
  • Rigorous quality control that includes GeneChip™ analysis
  • T7 MEGAscript™ IVT delivers up to 50,000-fold amplification
  • Single-tube format
  • Easy-to-use, high-recovery, magnetic-bead aRNA purification

Lowest RNA input requirements for a single round of amplification:

  • 20ng total RNA yields enough biotin-modified aRNA for most microarray formats, with results equivalent to those from samples prepared from significantly more starting material using other RT-IVT kits

Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, RNA Amplification

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type RNA Amplification Kit
Content And Storage • 110 μl First Strand Enzyme Mix (-20°C)
• 440 μl First Strand Buffer Mix (-20°C)
• 220 μl Second Strand Enzyme Mix (-20°C)
• 550 μl Second Strand Buffer Mix (-20°C)
• 660 μl T7 Enzyme Mix (-20°C)
• 2 × 1.1 ml T7 Biotin IVT Mix (-20°C)
• 10 μl Control RNA 1 mg/ml (-20°C)
• 1.75 ml Nuclease-free Water (any temperature)
• 6 ml aRNA Binding Buffer Concentrate (room temperature)
• 1.2 ml RNA Binding Beads (4°C)
• 30 ml aRNA Wash Solution Concentrate (room temperature)
• 15 ml aRNA Elution Solution (4°C or room temperature)
• 1 ml 5X Array Fragmentation Buffer (room temperature)
• 2 U-Bottom Plates (room temperature)
• 4 PCR Plates (room temperature)
• 1 Reservoir (room temperature)
• 10 mL Nuclease-free Water (any temperature)
Includes Label or Dye Yes
Labeling Method Direct Labeling
Reverse Transcriptase ArrayScript™
Sample Type Poly(A+) RNA, Total RNA
Format Kit
Product Line Ambion™, MessageAmp™ Premier
Quantity 100 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.