missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MessageAmp™ III RNA Amplification Kit

Product Code. 10448644
Change view
Click to view available options
Quantity:
10 Reactions
30 Reactions
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10448644 30 Reactions Each
10289484 10 Reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10448644 Supplier Invitrogen™ Supplier No. AM1793

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

The MessageAmp™ III aRNA Amplification Kit includes First-Strand Enzyme Mix, First-Strand Buffer Mix, Second-Strand Enzyme Mix, Second-Strand Buffer Mix, T7 Enzyme Mix, T7 Biotin IVT Mix, Control RNA (1mg/mL HeLa total RNA).

Provides extensive improvements over existing methods: fewer components, pipetting steps, and manipulations in the workflow

DNA microarrays are used to evaluate the expression levels of thousands of genes in a single experiment. High-density arrays are the method of choice for assessing gene expression on a genome-wide scale. One of the challenges associated with microarray technology, however, is that adequate amounts of labeled antisense RNA (known as cRNA or aRNA) are required. T7 linear amplification, commonly referred to as the Eberwine method, overcomes this challenge by simultaneously amplifying and labeling the RNA. This method is the basis of the MessageAmp™ III RNA Amplification Kit.

Workflow Improvements
The kit includes a set of master mixes, reducing the number of components, pipetting steps, and manipulations. Steps such as initial primer annealing and cDNA purification have been eliminated. All steps of the biotin labeling/amplification process take place in a single reaction tube for a simplified workflow.

  • Uses a glass fiber, filter-based aRNA purification method
  • Includes sufficient reagents for 10 reactions
  • All reactions occur in a single tube
  • Master Mix formulations reduce pipetting steps and hands-on time
  • Shortened processing time due to optimized incubations and condensed procedure
  • Less hands-on time means less opportunity to introduce errors
  • Start with as little as 20ng total RNA and produce enough aRNA for GeneChip™ analysis
  • Start with 100ng of total RNA (with a 4 hour IVT) and hybridize to a GeneChip; array in a single day
  • Shortens hands-on time, and reduces operator-dependant errors and variability

Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, RNA Amplification

TRUSTED_SUSTAINABILITY

Specifications

Product Type RNA Amplification Kit
Content And Storage The MessageAmp™ III aRNA Amplification Kit includes First-Strand Enzyme Mix, First-Strand Buffer Mix, Second-Strand Enzyme Mix, Second-Strand Buffer Mix, T7 Enzyme Mix, T7 Biotin IVT Mix, Control RNA (1 mg⁄mL HeLa total RNA) which are all stored at –20°C. Wash Buffer is stored at +4°C and aRNA Binding Buffer, Filter Cartridges, Collection Tubes, 8-Strip PCR Tubes & Caps (0.2 mL), 5X Array Fragmentation Buffer, and Nuclease-free Water are stored at room temperature.
Includes Label or Dye Yes
Labeling Method Direct Labeling
Reverse Transcriptase ArrayScript™
Sample Type Poly(A+) RNA, Total RNA
Format Kit
Product Line Ambion, MessageAmp
Quantity 30 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.