Learn More
Invitrogen™ Ambion™ MEGAshortscript™ T7 Transcription Kit
Brand: Invitrogen™ AM1354M
Code : 50
Additional Details : CAS Number : 7732-18-5 Weight : 0.07000kg
Specifications
T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-RNA 18S, TURBO DNase, Ammonium Acetate Stop Solution, and Gel Loading Buffer II are all stored at -20°C. Nuclease-free Water may be stored at any temperature. | |
T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-RNA 18S, TURBO DNase, Ammonium Acetate Stop Solution, Gel Loading Buffer II, and Nuclease-free Water | |
Kit with manual |
Gene Expression Analysis and Genotyping, in vitro Transcription | |
25 |