Learn More
Invitrogen™ Ambion™ MEGAclear™ Transcription Clean-Up Kit
Click to view available options
Quantity:
20 Preps
Unit Size:
Each
Specifications
Specifications
| For Use With (Application) | Gene Expression Analysis and Genotyping, in vitro Transcription |
| Includes | Binding Solution, Wash Solution Concentrate, 5M Ammonium Acetate, Elution Solution, Filter Cartridges, and Collection and Elution Tubes |
| Quantity | 20 Preps |