Learn More
Invitrogen™ Ambion™ MAXIscript™ T7 Transcription Kit
Product Code. 10440355
Change view
Click to view available options
Quantity:
30 Reactions
Unit Size:
Each
Product Code. Quantity unitSize
10440355 30 Reactions Each
1 options
Product Code. 10440355 Supplier Invitrogen™ Supplier No. AM1312M
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Content And Storage T7 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, and pTRI-b-actin are all stored at -20°C. Nuclease-free Water may be stored at any temperature.
For Use With (Application) DNA and RNA Purification and Analysis, Gene Expression Analysis and Genotyping, in vitro Transcription, Northern Blotting, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis and Blotting, Southern Blotting
Includes T7 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, pTRI-b-actin, and Nuclease-free Water
No. of Reactions 30
Product Type Kit with manual
Quantity 30 Reactions