missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAXIscript™ T7/T3 Transcription Kit

Product Code. 10331515
Change view
Click to view available options
Product Type:
T7/T3 In Vitro Transcription kit
No. of Reactions:
100 Reactions
30 Reactions
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type No. of Reactions unitSize
10331515 T7/T3 In Vitro Transcription kit 30 Reactions Each
10555785 T7/T3 In Vitro Transcription kit 100 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10331515 Supplier Invitrogen™ Supplier No. AM1324

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

T7 Enzyme Mix, T3 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, pTRI-b-actin, Nuclease-free Water

Fast synthesis of RNA probes for use in ribonuclease protection assays, in situ hybridization, and blot hybridizations

The Ambion MAXIscript T3/T7 In Vitro Transcription Kit synthesizes RNA probes in just 10 minutes for use in ribonuclease protection assays, in situ hybridization, and blot hybridizations. Each kit contains sufficient reagents for 15 reactions of each enzyme. MAXIscript Kits are very efficient at synthesizing full-length probes, even at limiting nucleotide concentrations, making them the perfect companion to ribonuclease protection assays. MAXIscript Kits are also an excellent choice for the synthesis of nonisotopically labeled probes. Labeling can be accomplished post-transcriptionally by using Ambion's BrightStar™ Psoralen-Biotin Labeling Kit, or during transcription by incorporating modified nucleotides into the reaction. MAXIscript Kits can also be used for the synthesis of modest amounts of unlabeled RNA (2–6μg/20μL reaction) of up to 2kb.

  • High yields of high specific activity, full-length RNA probes in just 10 minutes
  • Ideal probe synthesis kit for nuclease protection assays, Northern and Southern blots, and in situ hybridization
  • Includes TURBO DNase™
  • All kits contain quality tested DNase/RNase-free reagents
  • The MAXIscript In Vitro Transcription Kit synthesizes RNA probes with specific activities reaching 1 ×109 cpm/μg in just 10 minutes

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage T7 Enzyme Mix, T3 Enzyme Mix,10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, and pTRI-b-actin are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Final Product Type Synthetic mRNA, mRNA
Format Kit
Sample Type mRNA, Synthetic mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions 30 Reactions
Product Line Ambion, MAXIscript
Product Type T7/T3 In Vitro Transcription kit
Quantity 30 reactions
Promoter T3, T7
Shipping Condition Dry Ice
Yield 2-6 μg
Starting Material PCR Products, Linearized DNA plasmid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.