Learn More
Invitrogen™ MagMAX™-96 DNA Multi-Sample Kit
MagMAX™ DNA Multi-Sample 96-Well Kits are designed for rapid purification of high-quality genomic DNA from a variety of sample types including blood and blood cards, tissue, mouse tails, buccal swabs, buffy coats, and cultured cells.
£302.00 - £1242.00
Specifications
Format | Kit |
---|---|
Isolation Technology | Magnetic Bead |
Sample Type | Blood, Buccal Samples, Cells, Tissue |
Final Product Type | Genomic DNA |
For Use With (Application) | Genotyping, Real-Time Quantitative PCR (qPCR), PCR, Sequencing |
Includes:
- DNA Binding Beads, 10 mg/mL, 1 × 1.8mL (Same for both kits) Store at 2 to 8 °C
- DNA Elution Buffer 1 1 x 15mL (Same for both kits) Store at RT
- DNA Elution Buffer 2 1 x 15mL (Same for both kits)Store at RT
- Elution Plate, 96-well, 2 plates - Store at RT
- Wash Solution 1 Concentrate, 1 x 15mL for PN 4413021 - Store at RT
- Wash Solution 2 Concentrate, 1 x 50mL (Same for both kits) Store at RT
- Multi-Sample DNA Lysis Buffer, 1 x 55mL (Same for both kits) Store at RT
- MicroAmp Clear Adhesive Film, 1 x 4 films - Store at RT
- Nuclease-Free Water 1 x 50mL (Same for both kits) Store at RT
- PK Buffer 1 x 11mL (Same for both kits) Store at RT
- Processing Plate, 96-well, 1 plate - Store at RT
- Proteinase K, 1 x 850 μL - Store at -15 to -25 °C
- Ribonuclease A (RNase A), 1 x 530 μL - Store at -15 to -25 °C
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
12013729
|
Invitrogen™
4413021 |
96 Preps |
£302.00
96 preps |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11370912
|
Invitrogen™
4413022 |
480 Preps |
£1242.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MagMAX™ DNA Multi-Sample 96-Well Kits are designed for rapid purification of high-quality genomic DNA from a variety of sample types including blood and blood cards, tissue, mouse tails, buccal swabs, buffy coats, and cultured cells. These purification kits use MagMAX™ magnetic bead-based nucleic acid isolation technology to produce high yields of purified DNA, free from inhibitors that may affect downstream reactions such as genotyping assays, CE and next-generation sequencing, and high resolution melting analysis. The 96-prep and 5 x 96-prep kits are designed for higher throughput and automated format processing.
The demands for DNA yield and purity have become more stringent with the development of new analytical methods and technologies, such as high-throughput genotyping, and next-generation DNA sequencing platforms. MagMAX™ DNA Multi-Sample Kits meet the challenge by delivering highly pure DNA that is free of nucleases, proteins, and other inhibitors of downstream enzymatic reactions.
As shown in Table 1, DNA with high yield and purity can be obtained from a variety of materials using the MagMAX™ DNA Multi-Sample Kits. This is in contrast to filter membrane–based methods that often require different kit configurations for various input materials. For applications requiring maximal yields, for small batch sizes, or for DNA archival purposes MagMAX™ DNA Multi-Sample Kit (tube format, manual processing) may be desirable because larger sample inputs are accommodated. DNA yields suitable for most routine applications may be obtained using the MagMAX™-96 DNA Multi-Sample Kits (96-well plate format, compatible with automated handling) (Table 1).
The MagMAX™ DNA Multi-Sample Kit also delivers high yields of DNA from lipid or glycogen rich samples, such as brain and liver tissues, respectively. Figure 1 shows more than twice as much DNA was purified from tissues with the MagMAX™ DNA Multi-Sample Kit than with the leading competitor’s filter membrane–based method.
Most enzymatic molecular applications are prone to inhibition by analytes carried over from the sample matrix (e.g., heme, heparin, salts, protein) or the purification method itself (phenol, salts, alcohol). Figure 2 illustrates the effect of DNA purity on sensitive molecular applications, such as quantitative PCR (qPCR). DNA was extracted from blood using the MagMAX™-96 DNA Multi-Sample Kit, or a competitor’s filter membrane–based kit and subjected to real-time PCR analysis. The cycle threshold CT values obtained with the DNA purified using the MagMAX™ DNA Multi-Sample Kit were lower than those obtained with the filter membrane kit–purified DNA, indicating the MagMAX™ kit yields higher-purity DNA.
MagMAX™ DNA Multi-Sample Kits provide purified DNA that is suitable for a variety of Applied Biosystems application workflows (Figure 3), including SNP genotyping and DNA sequencing analysis (see Figure 4). To see the full performance description of DNA purified using the MagMAX™ DNA Multi-Sample Kits in TaqMan™ SNP genotyping assays and with BigDye™ chemistry in DNA sequence analysis, view the MagMAX™ DNA Multi-Sample Kit Product Bulletin.
Specifications
Kit | |
Blood, Buccal Samples, Cells, Tissue | |
Genotyping, Real-Time Quantitative PCR (qPCR), PCR, Sequencing | |
MagMAX™ | |
Genomic DNA |
Magnetic Bead | |
Genomic DNA | |
High-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols | |
DNA Multi-Sample Kit |
For Research Use Only. Not for use in diagnostic procedures.