missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ M-MLV Reverse Transcriptase
Description
Moloney Murine Leukemia Virus Reverse Transcriptase (M-MLV RT) is derived from a cloned region of the pol gene of M-MLV and isolated from an E. coli strain overexpressing this construct.
- In the presence of a single-stranded RNA or DNA template, primer, and deoxyribonucleotides, M-MLV Reverse Transcriptase (RT) will extend the oligonucleotide primer, producing a complementary strand
- Optimal reaction temperature: 37°C
- Final product size: 7kb or less
PCR & Real-Time PCR, Reverse Transcription
Order Info
Shipping Condition: Dry ice
Specifications
Specifications
| Quantity | 10,000 units |
| For Use With (Application) | RT-PCR |
| Sample Type | ssRNA, ssDNA, RNA-DNA Hybrid |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?