missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Ambion™ In Vivo Pre-Designed siRNA

Product Code. 15531114
Change view
Click to view available options
Quantity:
1 umol
250 nmol
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15531114 250 nmol Each
15541114 1 umol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15531114 Supplier Ambion™ Supplier No. 4457307
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Ambion™ In Vivo Pre-Designed siRNA

RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room temperature
Description Custom Ambion™ In-vivo Pre-designed siRNA, 250nmol, Solid, Powder, HPLC In-vivo Ready Purity, RNAi is the best Way to Effectively Knock Down Gene Expression to Study Protein Function in a Wide Range of Cell Types, Use our GeneAssist Custom siRNA Builder to Design and Order a Custom siRNA
Product Type Custom Ambion™ In-vivo Pre-designed siRNA
Purity HPLC In-vivo Ready
Quantity 250 nmol
Form Solid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.