missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ambion™ In Vivo Factor VII Positive Control siRNA

Product Code. 15581114
Change view
Click to view available options
Quantity:
250 nmol
50 nmol
Unit Size:
250nmol
50nmol
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15581114 50 nmol 50nmol
15501511 50 nmol Each
15561104 250 nmol 250nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15581114 Supplier Invitrogen™ Supplier No. 4459408
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ambion™ In Vivo siRNAs are designed using the proven Silencer™ Select algorithm and incorporate chemical modifications for superior serum stability with in vivo delivery.

Ambion™ In Vivo siRNA Controls

  • Validated siRNA controls for optimizing siRNA experiments targeting coding and noncoding transcripts
  • Positive Control siRNAs functionally tested in several common cell lines
  • Negative Controls functionally proven to have minimal effects on cell proliferation and viability
  • Include Silencer™ Select modifications for enhanced specificity or Ambion™ In Vivo modifications for increased stability
  • For use in mouse cells

What are Ambion™ In Vivo siRNAs?
Ambion™ In Vivo siRNAs are designed using the proven Silencer™ Select algorithm and incorporate chemical modifications for superior serum stability with in vivo delivery. The added serum stability does not compromise the performance of the siRNAs. Ambion™ In Vivo siRNAs have been shown to be non-toxic in vivo (mouse) and non-immunogenic (cell-based assays). In cell-based assays, Ambion™ In Vivo siRNAs exhibit equivalent or better potency than Silencer™ Select siRNAs. The result is better confidence in your in vivo RNAi experiments, better knockdown when paired with an efficient delivery solution, and the assurance of limited non-specific effects.

Factor VII (FVII) Positive Control siRNA
Ambion™ In Vivo Factor VII siRNA, In Vivo Ready, provides a positive control for experiments involving Ambion™ In Vivo siRNA delivery to mice. Factor VII (FVII/F7; also known as proconvertin) is a vitamin K-dependent serine protease that functions as a central protein in the coagulation cascade. FVII is synthesized in the liver and secreted to the plasma where it circulates in an inactive form. After delivery of FVII-targeting siRNA to the liver, knockdown can be measured by quantifying FVII protein levels in blood by robust, commercially available chromogenic assays. Moreover, blood samples can be collected from the same animal at multiple time points post-siRNA injection. These features enable much more rapid and efficient knockdown measurements for FVII than for gene targets that require quantitation of mRNA levels in liver tissue, making FVII an ideal control for in vivo siRNA delivery.

Quality Control
Ambion™ synthesizes and purifies each Ambion™ In Vivo siRNA in state-of-the-art facilities to meet the highest quality standards. As part of our rigorous quality control procedures, each RNA oligonucleotide is analyzed by MALDI-TOF mass spectrometry, and analytical HPLC is used to monitor purity. To provide the utmost in quality, we also assess each annealed siRNA by gel electrophoresis to confirm that the strands anneal properly. The result is premium-quality siRNA that is purified and ready to use.

In Vivo Ready Controls
Our 'In Vivo Ready' Control siRNAs are subjected to that extra level of purification and testing required for the introduction of siRNAs into animals. After HPLC purification and annealing, each siRNA is further purified utilizing a process that removes excess salt via a semi-permeable membrane. The result is highly pure siRNA with minimal salt content, suitable for in vivo applications. In vivo siRNAs are then sterile filtered, and tested for the presence of endotoxin. At concentrations of 50 μM in presence of deionized water, in vivo Ready siRNAs contain less than 5.0 mM Na+, less than 0.06 mM K+, and less than 0.02 mM Mg2+.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The siRNA (annealed) is shipped at ambient temperature as dried down oligonucleotides in a single tube, along with 1.75 mL Nuclease-free water for resuspension. Store at or below -20°C.
Description Ambion™ In-vivo Factor VII Positive Control siRNA, 50nmol, Unconjugated Dye, siRNA RNAi Type, Gene Knockdown Positive Control Type, Used for RNAi, Transfection, Factor VII Target Gene, –20°C Storage Temperature, For Research Use Only
Control Type Positive Control
RNAi Type siRNA
For Use With (Application) RNAi, Transfection
Includes The siRNA (annealed) is shipped at ambient temperature as dried down oligonucleotides in a single tube, along with 1.75mL nuclease-free water for resuspension
Label or Dye Unconjugated
Product Line Ambion
Product Type siRNA
Purity HPLC
Quantity 50 nmol
Shipping Condition Room Temperature
Target Factor VII
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.