missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Heat Seal Film for Sequencing & Fragment Analysis Sample Plates

Product Code. 15979177
Click to view available options
Quantity:
1 Pack
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15979177

Brand: Invitrogen™ 4337570

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Film for approximately 3500 seals.

Sample plate seal

This film is used to seal sample plates during sequencing and fragment analysis.

  • Used with MicroAmp™ Optical 384-Well Reaction Plate with Barcode, MicroAmp Optical 384-Well Reaction Plate with Barcode, MicroAmp Optical 96-Well Reaction Plate, MicroAmp Optical 96-Well Reaction Plate with Barcode

Sanger Sequencing, Sanger Sequencing Technology and Accessories, Sequencing

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Content And Storage Each package contains enough film for approximately 3,500 seals.
Plate Compatibility 384-Well Plates, 96-Well Plates
Quantity 1 Pack
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.