missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GTP Soln. (10 mM)

Product Code. 10698085
Click to view available options
Quantity:
100 μL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10698085

Brand: Invitrogen™ AM8130G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

This Invitrogen ribonucleotide triphosphate (rNTP) solution is a component of the MAXIscript™ Kits and is supplied in one tube containing 100 μL.

This Invitrogen ribonucleotide triphosphate (rNTP) solution is a component of the MAXIscript™ Kits (Cat. No. AM1312, AM1316, AM1314, AM1326, AM1324, AM1318, AM1322, AM1320, AM1310, and AM1308) and is supplied in one tube containing 100 μL. Each is subjected to RNase testing as well as a functional assay to ensure high levels of incorporation.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C.
Concentration 10 mM
For Use With (Application) In Vitro transcription
Label or Dye Unlabeled
Product Line Ambion
Quantity 100 μL
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.