missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gel Loading Solution (All-Purpose, Native Agarose)
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mL
missing translation for 'unitSize'
Each
Description
- Rigorously tested for non-specific endonuclease activity, exonuclease activity, RNase activity and for functionality
- Compatible with native agarose and polyacrylamide gels
- Bromophenol Blue and Xylene Cyanol dyes
- Supplied in one 1.4mL tube
- The same solution in Invitrogen kits
Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting
Informations sur la commande
Shipping Condition: Room Temperature
Spécification
Spécification
| Content And Storage | Store at -20°C. |
| Buffer | Sample Loading Buffers |
| Concentration | 10X |
| Label or Dye | Bromophenol Blue, Xylene Cyanol |
| Product Type | Gel Loading Solution |
| Gel Compatibility | Polyacrylamide Gels, Native Agarose |
| Quantity | 1 mL |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu