missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Formamide (Deionized)

Product Code. 10229524
Click to view available options
Quantity:
500 g
5 x 500 g
Unit Size:
2500g
500g
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10229524

Brand: Invitrogen™ AM9344

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Ambion formamide has been deionized, is nuclease-free, and comes packaged as a liquid in an amber glass bottle under argon.

Ambion formamide has been deionized, is nuclease-free, and comes packaged as a liquid in an amber glass bottle under argon. Supplied in 5 bottles containing 500 g each. Formamide lowers the melting temperature of nucleic acid duplexes and is widely used in applications such as Southern and northern blotting, nuclease protection assays, S1 mapping, and in situ hybridization. For critical applications, it is recommended that the formamide from previously opened bottles be deionized before use.

Appearance: Clear, colorless
Molecular Weight: 45.04
Purity: ≥99.5%
Quality Control: The product has been tested for RNase and DNase contamination.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Content And Storage Store at 4° or -20°C as small aliquots.
Packaging Type Bottle
Product Type Formamide
Product Line Ambion
Purity >99.5%
Quantity 5 x 500 g
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.