missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fast SYBR™ Green Cells-to-CT™ Kit
GREENER_CHOICE

Product Code. p-4918677
Click to view available options
No. of Reactions:
100 reactions
400 reactions
Unit Size:
100 reactions
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10678255

Brand: Invitrogen™ 4402956

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Ambion™ Fast SYBR™ Green Cells-to-CT™ Kit is a fast, easy, and robust method to go directly from cultured cells to real-time PCR results.

Ambion Fast SYBRGreen Cells-to-CT™ Kit is a fast, easy, and robust method to go directly from cultured cells to real-time PCR results. A breakthrough cell lysis and RNA stabilization technology eliminates the laborious and time-consuming RNA purification process completely. The kit includes sufficient reagents for 100 reactions. The Fast SYBRGreen Cells-to-CT Kit contains AmpliTaq™ Fast DNA Polymerase, UP, which allows instant hot start, thus minimizing non-specific product formation with fast real-time PCR.

  • Includes optimized reagents to work efficiently, right out of the box
  • Simple, effective Cells-to-CT technology enables sample preparation at room temperature in only 7 minutes, including DNase treatment
  • Validated accuracy, exceptional reproducibility, and maximal sensitivity from 10 to 100,000 cells per sample; results equivalent to those from purified RNA
  • 2-Step qRT-PCR

Extraordinary Value
The kit includes reagents for sample preparation integrated into a complete, optimized gene expression workflow including reverse transcription reagents and a high-performance PCR master mix with SYBRGreen.

Extraordinary Ease
The protocol begins with a simple 7-minute sample preparation procedure. First, cells are washed in PBS, and then lysed for 5 minutes at room temperature; Dnase treatment can be performed concurrently. The lysis reaction is stopped, and the sample is then ready for reverse transcription or storage at -20°C for up to 5 months. Because samples are processed directly in culture plates (96- or 384-well), sample handling and the potential for sample loss or transfer error are minimized, resulting in higher reproducibility. Following sample preparation, a portion of the cell lysate is added to an RT reaction followed by real-time PCR using the included master mix.

Extraordinary Performance
To demonstrate the performance of the kit, replicate experiments were run using either traditional RNA purification and real-time PCR analysis, or the Fast SYBRGreen Cells-to-CT Kit. The kit shows equivalent performance to results obtained using purified RNA for analysis of 10 to 100,000 cells. In addition, the sensitivity, efficiency, and dynamic range were shown to be superior to competitor lysate kits. Samples prepared with the kit have been tested for compatibility on various real-time PCR platforms including those from Applied Biosystems, Roche, and Bio-Rad.

Accessory Product:
The SYBRGreen Cells-to-CT Control Kit (Cat. No. 4402959) contains a Xeno™ RNA control and primer set and is recommended to monitor the efficiency of cell lysis and amplification inhibition in Cells-to-CT reactions.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20X
Content And Storage Store Stop Solution, DNase I and 20X RT Enzyme Mix at -20°C. Store Lysis Solution, 2X SYBR™ RT Buffer, Fast SYBR™ Green PCR Master Mix at 4°C. Store Nuclease-free Water at room temperature.
Detection Method SYBR
Polymerase AmpliTaq Fast DNA Polymerase
Reaction Speed Fast
For Use With (Equipment) 7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
Green Features Fewer resources used and less waste, Less hazardous
Label or Dye SYBR Green
Passive Reference Dye ROX (Pre-mixed)
Product Line Cells-to-CT, SYBR
Purification Time 5 min.
Purity or Quality Grade UP (Ultra Pure)
Quantity 100 reactions
Reverse Transcriptase M-MLV
Shipping Condition Dry Ice
For Use With (Application) RT-PCR
Hot Start Built-In Hot Start
No. of Reactions 100 reactions
Sample Type Cells
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.