missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ dNTPs for cDNA Probe Synthesis (10 mM)

Product Code. 10177724
Click to view available options
Quantity:
4 x 75ul
Unit Size:
300µL
This item is not returnable. View return policy

Product Code. 10177724

Brand: Invitrogen™ AM8200

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ideal for reverse transcription

Invitrogen dNTPs for cDNA Probe Synthesis (10mM) are the same high-quality deoxynucleotide triphosphate solutions provided in the RETROscript™ First Strand cDNA Synthesis Kit for RT-PCR (Cat. No. AM1710). They are subject to rigorous nuclease testing as well as a functional assay to ensure high levels of incorporation. Supplied as a set of 10mM solutions in four individual tubes containing 75μL each.

PCR and Real-Time PCR, Reverse Transcription

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in ultra-cold freezer (-68 to -85°C).
Concentration 10 mM
For Use With (Application) Reverse Transcription
Label or Dye Unlabeled
Product Line Ambion
Quantity 4 x 75ul
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.