Learn More
Invitrogen™ dNTP Mix (2.5 mM)
The same high-quality deoxynucleotide triphosphate solutions provided in the Invitrogen™ RETROscript™ First Strand cDNA Synthesis Kit for RT-PCR.
Brand: Invitrogen™ AM8228G
Code : 50
Additional Details : Weight : 0.00570kg
Description
The same high-quality deoxynucleotide triphosphate solutions provided in the Invitrogen™ RETROscript™ First Strand cDNA Synthesis Kit for RT-PCR (Cat. No. AM1710). They are subject to rigorous nuclease testing as well as a functional assay to ensure high levels of incorporation. Supplied in one tube containing 0.3 mL, premixed at a concentration of 2.5 mM each.
Order Info
Shipping Condition: Dry Ice
Specifications
Store at –20°C. | |
Reverse Transcription | |
Ambion™ | |
Dry Ice |
2.5 mM | |
Unlabeled | |
300 μL |
For Research Use Only. Not for use in diagnostic procedures.