missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DECAprime™ II DNA Labeling Kit

Product Code. 10391555
Change view
Click to view available options
Quantity:
100 Reactions
30 Reactions
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10391555 30 Reactions Each
10407304 100 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10391555 Supplier Invitrogen™ Supplier No. AM1455

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Exo– Klenow, 10X Decamer Solution, 5X Reaction Buffer minus dATP, 5X Reaction Buffer minus dCTP, DECAtemplate™ GAPDH-M, 0.5 M EDTA, nuclease-free water.

Technically superior to existing methods

The Ambion kit is for labeling DNA using a random-priming method, which has technical improvements over existing methods including the use of a high-purity, exonuclease-free Klenow and Random Decamers to produce probes with greater than 109 cpm/μg in 10 min reactions. The kit includes sufficient reagents for 30 reactions.

Don't Trade Specific Activity for Yield

There is a trade-off between probe yield and probe specific activity when using the random-priming method for labeling DNA. Both limiting nucleotide and template mass affect probe yield. Until the labeled nucleotide becomes limiting, the larger the amount of DNA template used, the greater the yield of probe. However, once the labeled nucleotide becomes limiting, additional template will only result in lower specific activity, since the unlabeled template competes with the labeled probe for target.

Maximal Yields of High Specific Activity Probes
The DECAprime™ II DNA Labeling Kit produces probes with maximum specific activity even when the DNA template is impure or the quantity is unknown or very low. Data shows that low template amounts require long incubation times to reach maximum specific activity. Under these conditions, extended reaction times will increase both the yield and the specific activity of the probe.

Accessory Products:
The NucAway™ Spin Columns (Cat. No. AM10070) are recommended for probe purification.

  • Fast, 10 min reaction time
  • Elimination of Klenow exonuclease activity maximizes specific activity
  • Improved labeling kinetics maximizes yields
  • Flexible: both -dATP and -dCTP buffers supplied with each kit
  • Decamers produce greater primer-template stability

DNA and RNA Purification and Analysis, DNA Labeling, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting, Nucleic Acid Labeling and Oligo Synthesis, Southern Blotting

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type DNA Labeling Kit
Detection Method Radioactive
Content And Storage Exo– Klenow, 10X Decamer Solution, 5X Reaction Buffer minus dATP, 5X Reaction Buffer minus dCTP, DECAtemplate™ GAPDH-M, and 0.5 M EDTA should be stored at
–20°C. Nuclease-free water may be stored at any temperature.
Final Product Type Probes (Labeled DNA)
Label or Dye 32P (Phosphorous-32)
Includes Label or Dye No
Labeling Target DNA (General)
Labeling Method Direct Labeling
Shipping Condition Dry Ice
Format Kit
Product Line Ambion, DECAprime
Quantity 30 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.