missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Decade™ Markers System (to make radiolabeled RNA markers)

Product Code. 10299104
Change view
Click to view available options
Quantity:
10 reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10299104 10 reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10299104 Supplier Invitrogen™ Supplier No. AM7778

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Decade™marker RNA, Polynucleotide Kinase, Kinase Reaction Buffer, Cleavage Reagent, Gel Loading Buffer II, and Nuclease-free water

For making radiolabeled RNA markers

The Invitrogen™ Decade™ Marker System produces a set of radiolabeled RNA molecules of 150, 100, 90, 80, 70, 60, 50, 40, 30, 20, and 10 nucleotides. The molecular weight markers are ideally suited for experiments where RNA molecules less than 150 nucleotides in length are being studied. Sufficient reagents are provided for 10 marker preparation reactions. This marker set is particularly useful for research laboratories studying tRNA, snRNA, snoRNA, miRNA, siRNA and other small RNA molecules. The system includes all of the reagents necessary to produce the molecular weight markers except gamma-32P-ATP. The Decade™ Markers derive from a single, gel-purified transcript that is provided. The transcript is 5' end-labeled using the kinase and kinase buffer supplied with the kit. The kinase reaction is then diluted into a cleavage reagent that generates the molecular weight marker set in a five minute, room temperature reaction. Gel loading buffer from the kit is then added and the marker is ready for electrophoresis. The labeling, cleavage, and gel preparation steps can be completed in less than an hour. Compatible with denaturing polyacrylamide gels.

Acrylamide Gel Electrophoresis, DNA and RNA Purification and Analysis, Northern Blotting, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Decade™ Marker RNA should be stored at below -70°C. Polynucleotide Kinase, Kinase Reaction Buffer, Cleavage Reagent, Gel Loading Buffer II are stored at 4°C or -20°C. Nuclease-free water is stored at any temperature.
Concentration 100 μg/ml
Gel Type Polyacrylamide
Ready to Load No
Size Range 10 to 150 bp
Gel Compatibility Denaturing Polyacrylamide Gels
Quantity 10 reactions
Shipping Condition Dry Ice
Product Line Ambion, Decade
Product Type Markers System

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.