missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellsDirect Resuspension & Lysis Buffers includes 10 ml Resuspension Buffer & 1 ml Lysis Buffer

Product Code. 10666713
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10666713 1 kit 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10666713 Supplier Invitrogen™ Supplier No. 11739010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

10mL Resuspension Buffer, 1mL Lysis Buffer

These two buffers are to be used only as a complement to the SuperScript™ III Platinum™ CellsDirect Kits when large number of samples are processed.

These two buffers are to be used only as a complement to the SuperScript™ III Platinum™ CellsDirect Kits when large number of samples are processed. See manual of SuperScript III Platinum CellsDirect Kits for protocol recommendations.

  • Fewer resources used, less hazardous, less waste

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Store in freezer (-5 to -30°C).
Product Line CellsDirect
Product Type Cell Lysis Buffer
Quantity 1 kit
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.