missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATP Solution (10 mM)

Product Code. 10688085
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
Each
Product Code. Quantity unitSize
10688085 100 μL Each
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10688085

Brand: Invitrogen™ AM8110G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

This Invitrogen ribonucleotide triphosphate (rNTP) solution is a component of the MAXIscript™ Kits and is supplied in one tube containing 100 μL.

This Invitrogen ribonucleotide triphosphate (rNTP) solution is a component of the MAXIscript™ Kits (Cat. No. AM1312, AM1316, AM1314, AM1326, AM1324, AM1318, AM1322, AM1320, AM1310, and AM1308) and is supplied in one tube containing 100 μL. Each is subjected to RNase testing as well as a functional assay to ensure high levels of incorporation. It may also be used in ligation and phosphorylation reactions.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C.
Concentration 10 mM
For Use With (Application) In Vitro transcription
Labeling Method RNA Labeling
Label or Dye Unlabeled
Product Line Ambion
Quantity 100 μL
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.