missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ambion™ ANTI-RNase (15-30U/μL)
Click to view available options
Quantity:
10,000 U
Unit Size:
Each
Specifications
Specifications
| Concentration | 15 to 30U/μL |
| Content And Storage | Store at -20°C. Do not store in a frost–free freezer or at -70°C. |
| For Use With (Application) | Non-covalently binds and inactivates neutral pancreatic RNase A–type enzymes |
| Product Type | ANTI-RNase |
| Quantity | 10,000 U |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction