missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Acrylamide/Bis 19:1, 40% (w/v) solution

Product Code. 10298364
Change view
Click to view available options
Quantity:
500 mL
2 x 500 mL
Unit Size:
Each
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10298364 500 mL Each
10695785 2 x 500 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10298364 Supplier Invitrogen™ Supplier No. AM9022

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ambion Acrylamide/Bis 19:1 is a 40% (w/v) solution of acrylamide (38%) and bis-acrylamide (2%) ideal for use in ribonuclease protection assay, sequencing gels, and sizing DNA or RNA fragments.

Ambion Acrylamide/Bis 19:1 is a 40% (w/v) solution of acrylamide (38%) and bis-acrylamide (2%) ideal for use in ribonuclease protection assay, sequencing gels, and sizing DNA or RNA fragments. The solution is provided in a ready-to-use form, reducing the dust, inhalation, and contact hazard associated with weighing and preparing acrylamide and bis-acrylamide powders and solutions. Concentrations are carefully controlled and all solutions are micro-filtered prior to packaging. All Ambion acrylamides are rigorously tested for contaminating non-specific endonuclease, exonuclease, and RNase activity, and for functionality.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C.
Format Bottle
Form Liquid
Quantity 500 mL
Shipping Condition Room Temperature
Product Line Ambion

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.