Learn More
Specac™ Ambient Temperature Cell Holder
Product Code. 15210248
Encompass_Preferred
Click to view available options
Type:
Ambient Temperature Cell Holder
Product Code. 15210248
Brand: Specac™ GS20740
This cell holder has been designed to hold a variety of liquid and solid sample cell holders at ambient temperatures and be can slotted directly into any FTIR instrument.
Specifications
Type Ambient Temperature Cell Holder