missing translation for 'onlineSavingsMsg'
Learn More

Ambersep™ 900(OH), ion exchange resin

Product Code. 11339575
Click to view available options
Quantity:
100 g
500 g
1000 g
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 11339575

Brand: Thermo Scientific Alfa Aesar L19035.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9017-79-2

Specially sized for use in Ambersep resin systems and high-flow rate condensate polishing. It holds regenerable mixed bed applications such as the production of ultrapure water for semiconductor manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Specially sized for use in Ambersep resin systems and high-flow rate condensate polishing. It holds regenerable mixed bed applications such as the production of ultrapure water for semiconductor manufacturing.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9017-79-2
MDL Number MFCD00802695

Specifications

Odor Odorless
Quantity 1000 g
Solubility Information Insoluble in water.
Chemical Name or Material Ambersep™ 900(OH), ion exchange resin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.