missing translation for 'onlineSavingsMsg'
Learn More

Amberlyst™ A-26(OH), ion exchange resin

Product Code. 11473597
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473597 1000 g 1000g
11483597 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473597 Supplier Thermo Scientific Alfa Aesar Supplier No. A17361.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39339-85-0

Amberlyst™ A-26(OH) is widely utilized to catalyze reactions such as aldol condensation and carbonylation. It is used to remove the anionic transition metal complexes from aqueous and non-aqueous mediums, acids and mercaptans from hydrocarbons and other non-polar solvents. It plays an essential role for the removal of oleic acid from chlorinated hydrocarbons and acids from phenol-acetone solutions. It is actively involved in the determination of cadmium by flow injection atomic absorption spectrometry and as packaging material for column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlyst|r A-26(OH) is widely utilized to catalyze reactions such as aldol condensation and carbonylation. It is used to remove the anionic transition metal complexes from aqueous and non-aqueous mediums, acids and mercaptans from hydrocarbons and other non-polar solvents. It plays an essential role for the removal of oleic acid from chlorinated hydrocarbons and acids from phenol-acetone solutions. It is actively involved in the determination of cadmium by flow injection atomic absorption spectrometry and as packaging material for column chromatography.

Solubility
Insoluble in water.

Notes
Exposure to air absorbs carbon dioxide. Incompatible with strong acids, bases and oxidants.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39339-85-0
MDL Number MFCD00145579

Specifications

Quantity 1000 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlyst™ A-26(OH), ion exchange resin
compliance-icons
Product Identifier
  • Amberlite A-26(OH), ion exchange resin
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Amberlyst A 26

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.