missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ XAD™-2, TRC

Product Code. 30322618
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322618 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322618 Supplier Toronto Research Chemicals Supplier No. TRCA57649825G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Amberlite XAD-2
CAS 9003-70-7
Molecular Formula [CH 2 CH(C 6 H 5 )]x[CH 2 CH[C 6 H 4 (CHCH 2 )]]y
Quantity 25 g
Synonym Bio-Beads S, Bio-Beads SM 2, Bio-Beads SX 12, Bio-Beads SX 2, Bio-Beads SX 3, Bio-Beads SX 4, Bio-Beads SX 8, Bio-Rad SM 2, Bonopore 7, CDR 10, CS 1, CS 1 (catalyst support), CSPO, Cekachrom 2, Cekachrom 3, Chemisnow SGP 150C, Chromosorb 101, Chromosorb 102, D 001X14, D 005, D 005 (catalyst), DVB 960-styrene copolymer, DVBS, Diethenylbenzene-ethenylbenzene polymer, Divinylbenzene-ethylenediamine-formaldehyde-phosphoric acid-styrene copolymer, Divinylbenzene-polystyrene copolymer, Divinylbenzene-styrene copolymer, Divinylbenzene-styrene polymer, Divinylbenzene-styrene resins, Divinylbenzene-vinylbenzene copolymer, Finepearl 3000F, GDX 101, GS 0605, Ganz Pearl GS 0605, Ganzpearl GS 1105, Gaskuropack 54, HAP 1, HP 01, HP 01 (vinyl polymer), HP 60, HY 1, HY 1 (adsorbent), HZ-PS 20SS, HZ-PS 816, HZ-PS 818, HZ-PS 830, Hamilton PRP 1, Hitachi 3010, InertSep RP 1, JSR-SX 8705P01, JSR-SX 8742-08, JSR-SX 8743, JSR-SX 8743-05, JSR-SX 8743-08, JSR-SX 8743C03, KPN 258, MCT 1, MOD 40/100, MR 500, NGW 04, Nanotex PF 032A, Nanotex PF 082A, Novabiochem 01-64-0022, Oligo R 3, P 8D, PLRP-S, PRLP-S, PRP 1, PS 1/85, PS 10V, PS 10t100, PS 20t100, PS 40t100, Poly(divinylbenzene-styrene), Poly(styrene-co-DVB), Poly(styrene-co-divinylbenzene), Poly(styrene-divinylbenzene), PolyVVFE, Polyacene SD, Polyhipe X 20PV90, Polysorb 1, Polysorb 40/100, Polysorb 4T, Polysorb C, Polysorb C (copolymer), Polysorb S 40/100, Polysorb S 60/100, Polyspher PST 10, Polystyrene-DVB, Polystyrene-divinylbenzene copolymer, Polystyrene-divinylbenzene polymer, Pontybond, Poros 10R2, Poros 20R1, Poros R 1, Poros R 1-50, Poros R 2Purasorb AP 250, Rexolite, Rexolite 1422, Rexolite 2200, Rexolite I, Rexolite II, SD 15H60, SDVB, SDX 5, SGP 150C, SGP 50C, SGP 70C, SX 713, SX 8742-08, Shodex 801, Source, Sprinbeads SN 110, Staphyloid GS 30A, Staphyloid GS 60A, Styragel, Styrene copolymer with divinylbenzene, Styrene-DVB copolymer, Styrene-divinybenzene copolymer, Styrene-divinylbenzene copolymer, Styrene-divinylbenzene polymer, Styrene-divinylbenzene resin, Styrene-divinylbenzene-2,2'-bipyridyl copolymer, Sunpak A, Synachrom, TSK G4000H, TVEKS, TVEX, TX-ER 6, Vinylsorb S, WD 4, WDX 3, XE 53BNGF, Y 29/11
InChI Formula InChI=1S/C8H8.C6H6.2C3H6/c1-2-8-6-4-3-5-7-8;1-2-4-6-5-3-1;2*1-3-2/h2-7H,1H2;1-6H;2*3H,1H2,2H3
SMILES CC=C.CC=C.C=Cc1ccccc1.c2ccccc2
IUPAC Name benzene;prop-1-ene;styrene
Formula Weight 266.2035
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.