missing translation for 'onlineSavingsMsg'
Learn More

Amberlite XAD-1180

Product Code. 11382887
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382887 500 g 500g
11372887 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382887 Supplier Thermo Scientific Alfa Aesar Supplier No. L16097.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97396-56-0

Polymeric adsorbent for hydrophobic compounds: antibiotics, vitamins, amino acids, enzymes; for purification: gallium, myxovirescins, phosphoric acid, riboflavin salts. Amberlite™ XAD1180N can be used as a polymeric adsorbent. The regeneration of this adsorbent has been effective in alkaline medium and tests have shown the adsorbent efficiently removes chromium from tannery waste water. Mainly used in the Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polymeric adsorbent for hydrophobic compounds: antibiotics, vitamins, amino acids, enzymes; for purification: gallium, myxovirescins, phosphoric acid, riboflavin salts. Amberlite™ XAD1180N can be used as a polymeric adsorbent. The regeneration of this adsorbent has been effective in alkaline medium and tests have shown the adsorbent efficiently removes chromium from tannery waste water. Mainly used in the Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97396-56-0
MDL Number MFCD00145830

Specifications

Merck Index 14,382
Quantity 500 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlite XAD-1180

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.