missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ XAD-1180, polymeric adsorbent

Product Code. 10277600 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10277600 500 g Plastic Bottle 500g
10003400 100 g Plastic Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10277600 Supplier Thermo Scientific Acros Supplier No. 299155000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9003-69-4 | C10H10

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-69-4
Molecular Formula C10H10
Molecular Weight (g/mol) 0.00
MDL Number MFCD00132704
InChI Key MYRTYDVEIRVNKP-UHFFFAOYSA-N
Synonym divinylbenzene, 1,2-divinylbenzene, o-divinylbenzene, benzene, diethenyl, benzene, 1,2-diethenyl, divinylbenzen, divinyl benzen, divinyl-benzene, poly divinylbenzene, divinylbenzene dvb
PubChem CID 66666
SMILES *

Specifications

Physical Form Beads
Particle Size Particle size analysis: uniformity coefficient: <1.80, harmonic mean size: 0.350 to 0.600mm, <0.250mm: <5.0%,>0.850mm: <10.0%
Packaging Plastic Bottle
Water Retention Capacity moisture holding capacity: 61.0 to 67.0%
Color White to Yellow
Merck Index 14, 382
Quantity 500 g
IUPAC Name 1,2-bis(ethenyl)benzene
Chemical Name or Material Amberlite™ XAD 1180

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.