missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRN-78, OH-form, ion-exchange resin

Product Code. p-4089377 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10179679

Brand: Thermo Scientific Acros 429291000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 11128-95-3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11128-95-3
MDL Number MFCD00145822

Specifications

Physical Form Solution
Packaging Plastic bottle
Color Colorless to Yellow
Quantity 100 g
Chemical Name or Material Amberlite™ IRN-78 ion-exchange resin, Ion-exchange Resin, OH-form
compliance-icons
Product Identifier
  • Amberlite(TM) IRN78 resin, hydroxide form
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.