missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRN-78, ion exchange resin, nuclear grade

Product Code. p-4824597
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 15437015

Brand: Thermo Scientific Alfa Aesar 045591.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Strongly basic anion exchange resin | CAS: 11128-95-3 | Styrene-DVB

Amberlite™ IRN-78 is widely employed in removing radioactive anions for example 131 Iodine and 133 Iodine from waste systems. It also removes the traces of chloride contamination from reactor coolant systems and controls the boron level in the primary system. At higher pH conditions, it is used to remove anionic impurities from secondary streams in the presence of ammonia, morpholine or other amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11128-95-3
Molecular Formula Styrene-DVB
MDL Number MFCD00145822

Specifications

Physical Form Beads
Color Amber
Quantity 500 g
Solubility Information Insoluble in water.
Grade Nuclear
Chemical Name or Material Amberlite™ IRN-78
compliance-icons
Product Identifier
  • Amberlite IRN-78, ion exchange resin, nuclear grade
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : amberlite IRN 78

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.