missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRC120 Na Ion Exchange Resin

Product Code. 11358206
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358206 500 g 500g
11348206 100 g 100g
11368206 2500 g 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11358206 Supplier Thermo Scientific Alfa Aesar Supplier No. L15199.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 65098-55-7 | C13H10ClNO4S | 311.736 g/mol

Amberlite™ IR-120(Na) ion exchange resin is used in water softening, dealkalinization and deionization. It is used for the recovery of toxic metals. It is also used in separation medium for TLC, paper chromatography and LPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications

Amberlite™ IRC120 Na Ion Exchange Resin is used in water softening, dealkalinization and deionization. It is used for the recovery of toxic metals. It is also used in separation medium for TLC, paper chromatography and LPLC.

Solubility

Insoluble in water.

Notes

Water softening, dealkalinization, deionization

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65098-55-7
Molecular Formula C13H10ClNO4S
Molecular Weight (g/mol) 311.736
MDL Number MFCD00132707
InChI Key APBOVLPLJFJSRI-UHFFFAOYSA-N
Synonym 3-3-chlorophenylsulfonamido benzoic acid, 3-3-chloro-benzenesulfonylamino-benzoic acid, 3-3-chlorophenyl sulfonyl amino benzoic acid, benzoicacid, 3-3-chlorophenyl sulfonyl amino, 3-3-chlorobenzenesulfonamido benzoic acid, amberlite ir-120, 3-3-chlorophenyl sulfonylamino benzoic acid, 3-3-chlorophenyl sulfonamido benzoic acid
PubChem CID 8190984
IUPAC Name 3-[(3-chlorophenyl)sulfonylamino]benzoic acid
SMILES C1=CC(=CC(=C1)NS(=O)(=O)C2=CC(=CC=C2)Cl)C(=O)O

Specifications

Merck Index 14,382
Quantity 500 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlite™ IRC120 Na Ion Exchange Resin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.