missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRA-958(Cl), ion exchange resin

Product Code. 11325457
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11325457 1 kg 1kg
11335457 250 g 250g
11315457 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11325457 Supplier Thermo Scientific Alfa Aesar Supplier No. 042702.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Removes Color, organics, and Cyanide

Removal of color, organics, and cyanideAmberlite™ IRA-958(Cl), ion exchange resin is used as an organic scavenger for the adsorption of natural organic matter from surface water. It is an acrylic resin for decolorization and the removal of organic material. Further, it is useful for decolorizing sugar juices and cane syrups, citric acid purification and capturing metal complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Amberlite IRA-958(Cl)
Physical Form Beads
MDL Number MFCD00211600
Color White
Quantity 1 kg
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.