missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRA-900, Cl-form, ion-exchange resin

Product Code. 10226260 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10226260 500 g Plastic bottle 500g
10321004 100 g Plastic bottle 100g
13458448 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10226260 Supplier Thermo Scientific Acros Supplier No. 202315000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9050-97-9 | C4H12ClN | 109.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9050-97-9
Molecular Formula C4H12ClN
Molecular Weight (g/mol) 109.60
MDL Number MFCD00132712
InChI Key OKIZCWYLBDKLSU-UHFFFAOYSA-M
Synonym tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl
PubChem CID 6379
ChEBI CHEBI:7070
SMILES [Cl-].C[N+](C)(C)C

Specifications

Physical Form Beads
Particle Size Particle size analysis : uniformity coefficient: <1.8, harmonic mean size: 0.650 to 0.820mm, <0.300mm: <0.5%,> 1.180mm: <3.0%
Packaging Plastic bottle
Volume Total Capacity (Cl-):> 1.0 eq/L
Water Retention Capacity (Cl-): 58 to 64%
Fieser 01,511; 02,267; 04,267; 06,302; 07,182
Color Beige/Brown to Ivory
Quantity 500 g
IUPAC Name tetramethylazanium;chloride
Chemical Name or Material Amberlite™ IRA-900

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.