missing translation for 'onlineSavingsMsg'
Learn More

Amberlite IRA-402(OH), ion exchange resin, Thermo Scientific™

Product Code. 11463707
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463707 500 g 500g
11453707 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463707 Supplier Thermo Scientific Alfa Aesar Supplier No. A18185.36

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Amberlite IRA-402(OH) ion exchange resin is a strongly basic gel-type anion exchange resin. It is used to adsorb uranium(VI) from sulphate, removal of cyanide and zinc cyanide. It is also used to produce perchlorate and the free nucleoside. Further, it is used for water conditioning.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52439-77-7
MDL Number MFCD00145564

Specifications

Density 0.656
Merck Index 14,382
Quantity 500 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlite IRA-402(OH), ion exchange resin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.