missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amberlite™ IR-120, Cation Exchange Resin, for Ion Chromatography, gel type (Sodium form, strongly acidic), Fisher Chemical
Shop All Fisher Chemical ProductsSpecifications
Specifications
| Physical Form | Solid |
| Packaging | Plastic powder jar |
| MDL Number | 132707 |
| Quantity | 500 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?