missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ 1000H Cation, for Ion Chromatography, Gel, Strongly Acidic, Hydrogen form (size 0.62-0.80mm), Fisher Chemical

Product Code. 11907001 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11907001 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11907001 Supplier Fisher Chemical Supplier No. IE/1012/53
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Physical Form Solid
CAS 69011-20-7
Melting Point 0°C
Boiling Point 100°C
Packaging Plastic powder jar
Molecular Formula C28H30
MDL Number MFCD00132722
Color Amber
Synonym porapak p, lewatit c267 ion exchange resin, ion exchange resin, benzene, diethenyl-, polymer with ethenylbenzene and ethenylethylbenzene, cation exchange resin, ethidium bromide adsorber, divinylbenzene, ethylvinylbenzene, vinylbenzene copolymer, ethenylbenzene, diethenylbenzene, ethylethenylbenzene polymer, ethylbenzene, ethylethenylbenzene, diethenylbenzene polymer, sulfonated, benzene, diethenyl-, polymer with ethenylbenzene and ethenylethylbenzene, sulfonated
Quantity 500 g
InChI Key NWUYHJFMYQTDRP-UHFFFAOYSA-N
SMILES *
IUPAC Name 1,2-bis(ethenyl)benzene;1-ethenyl-2-ethylbenzene;styrene
Molecular Weight (g/mol) 0.00
PubChem CID 172244
Show More Show Less
compliance-icons
Product Identifier
  • Amberjet 1000 H Resin
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Benzene, diethynyl-,polymer with ethenylbenzene and ethenylethylbenzene, sulfonated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.