missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom™, 50WX8, 200-400 mesh, H-form, Thermo Scientific™

Product Code. 17565605 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.50kg
500g
This item is not returnable. View return policy

Product Code. 17565605

Brand: Thermo Scientific Acros 472935000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 69011-20-7, 7732-18-5
Molecular Formula C28H30
Molecular Weight (g/mol) 0.00
MDL Number MFCD00132722
InChI Key NWUYHJFMYQTDRP-UHFFFAOYSA-N
IUPAC Name 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene
SMILES *

Specifications

Physical Form Beads
CAS Min % 42%, 50%
CAS Max % 50%, 58%
Ion Exchange Capacity ≥1.7 eq/L (H form)
Particle Size 200 to 400 mesh, ≤15 % (< 44 μm), ≤15 % (> 150 μm)
Packaging Plastic Bottle
Water Retention Capacity 50 to 58 % (H form)
Color Beige to Green/Beige to Light Brown
Quantity 500 g
Solubility Information Insoluble in water
Chemical Name or Material AmberChrom™
Show More Show Less
compliance-icons
Product Identifier
  • AmberChrom 50WX8, H-form
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Styrene, Divinylbenzene and EthylstyreneCopolymer, Sulfonated
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.