missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom™, 50WX8, 100-200 mesh, H-form, Thermo Scientific™

Product Code. 17535605 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17535605 500 g 500g
17525605 100 g 100g
17515605 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17535605 Supplier Thermo Scientific Acros Supplier No. 472925000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 69011-20-7 , 7732-18-5
Molecular Formula C28H30
Molecular Weight (g/mol) 0.00
MDL Number MFCD00132722
InChI Key NWUYHJFMYQTDRP-UHFFFAOYSA-N
IUPAC Name 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene
SMILES *

Specifications

Physical Form Beads
CAS Min % 42%, 50%
CAS Max % 50%, 58%
Ion Exchange Capacity ≥1.7 eq/L (H form)
Particle Size 100 to 200 mesh, ≤15 % (< 88 μm), ≤15 % (> 250 μm)
Packaging Plastic Bottle
Water Retention Capacity 50 to 58 % (H form)
Color Beige to Green/Beige to Light Brown
Quantity 500 g
Solubility Information Insoluble in water
Chemical Name or Material AmberChrom™
Show More Show Less
compliance-icons
Product Identifier
  • AmberChrom 50WX8, H-form
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Styrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.