missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom™, 1x8, 50-100 mesh, Cl-form, Thermo Scientific™

Product Code. 17505615 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 17505615

Brand: Thermo Scientific Acros 472950025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 69011-19-4, 7732-18-5
Molecular Formula C31H39N
Molecular Weight (g/mol) 0.00
MDL Number MFCD00146763
InChI Key HADXLRSCRPYPJJ-UHFFFAOYSA-N
IUPAC Name 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene; trimethylamine
SMILES *

Specifications

Physical Form Beads
CAS Min % 35%, 55%
CAS Max % 45%, 65%
Ion Exchange Capacity ≥1.2 eq/L (Cl form)
Particle Size 50 to 100 mesh, ≤15 % (< 180 μm), ≤15 % (> 500 μm)
Packaging Plastic Bottle
Water Retention Capacity 43 to 48 % (Cl form)
Color Tan to Yellow to White
Quantity 2.5 kg
Solubility Information Insoluble in water
Chemical Name or Material AmberChrom™
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.