Learn More
BRAND™ Amber LDPE Amber Bottle
Product Code. p-7164760 Shop All Brand Products
Click to view available options
Capacity (Metric):
50 mL
250 mL
500 mL
Unit Size:
Each
Product Code. 10129810
Brand: BRAND™ 130228
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Amber LDPE Amber Bottle features a narrow mouth and screw cap closure.
  • Opaque
  • For storage of light-sensitive substances
  • With screw cap
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50 mL
Color Amber
Material LDPE
Thread Size (Metric) GL 18
Mouth Narrow
Closure Type Screw Cap