Learn More
BRAND™ Amber 3.3 Borosilicate Glass Volumetric Flask With Polypropylene Stoppers
Amber glass to protect light-sensitive contents.
Brand:  BRAND™ 37489
63.44 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.17960kg
 View more versions of this product
Product Code. 15683967
  • £80.85 / Pack of 2
In Stock
to see stock.
Add to basket
Description
Description
  • Manufactured from Boro 3.3 glass
  • Acc. to DIN EN ISO 1042
  • Includes USP batch certificate
  • Class A, DE-M marking
Specifications
Specifications
PP (Polypropylene)
Borosilicate Glass (3.3)
A
250 mL
Volumetric flask
Stopper