Learn More
Emma Safety Footwear Amazone XXD Safety Shoes
Product Code. 15905160
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. 15905160
Brand: Emma Safety Footwear MM33603644
Only null left
Add to basket
Add to basket
A safe, well protected shoe with a sturdy steel toe. The Amazone's XXD's 'sticky grip' nitrile rubber, Vibram designed sole is highly resistant to extreme temperatures and corrosive chemicals.
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck
  • Heat Resistant Outsole
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Black
Design Amazone XXD
Liner Material Hydro-Tec™ Sanitized Silver
Material Oiled Nubuck
Size 44
Toe Material Steel